2,4,6-Collidine 98%

Supplier: Ambeed
Danger

A208876-10G A208876-25G
77030-444EA 5.31 USD
77030-444 77030-446
2,4,6-Collidine 98%
2,4,6-Collidine

Precaución: for Research Use Only

Formula: C₈H₁₁N
MW: 121.18 g/mol
MDL Number: MFCD00006338
CAS Number: 108-75-8
UN: 1992
ADR: 3,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR