p-Xylenolphthalein 97%

Supplier: Ambeed
Warning

Synonyms: 3,3-Bis(4-hydroxy-2,5-dimethylphenyl)isobenzofuran-1(3H)-one

A208819-100G A208819-25G
77030-434EA 64.72 USD
77030-434 77030-436
p-Xylenolphthalein 97%
p-Xylenolphthalein

Precaución: for Research Use Only

Formula: C₂₄H₂₂O₄
MW: 374.44 g/mol
MDL Number: MFCD00042785
CAS Number: 50984-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR