2,5-Dichloropyridine 98%

Supplier: Ambeed
Warning

A208765-100G A208765-25G A208765-500G
76999-554EA 42 USD
76999-554 76999-556 76999-558 76999-560
2,5-Dichloropyridine 98%
2,5-Dichloropyridine

Precaución: for Research Use Only

Formula: C₅H₃Cl₂N
MDL Number: MFCD00006239
CAS Number: 16110-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR