7-Bromo-2-methylbenzoxazole 97%

Supplier: Ambeed
Warning

A208674-100MG A208674-250MG A208674-1G
77030-428EA 6 USD
77030-428 77030-430 77667-752
7-Bromo-2-methylbenzoxazole 97%
7-Bromo-2-methylbenzoxazole

Precaución: for Research Use Only

Formula: C₈H₆BrNO
MDL Number: MFCD12965124
CAS Number: 1239489-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR