3-Bromo-6-iodoquinoline 98%

Supplier: Ambeed
Warning

A208467-100MG A208467-250MG
76986-640EA 68.96 USD
76986-640 76986-642
3-Bromo-6-iodoquinoline 98%
3-Bromo-6-iodoquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrIN
MW: 333.95 g/mol
MDL Number: MFCD12024485
CAS Number: 205114-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR