(5-Fluoro-2-(trifluoromethyl)phenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(5-Fluoro-2-(trifluoromethyl)phenyl)acetic acid

A208081-10G A208081-1G A208081-250MG
77030-400EA 136.86 USD
77030-400 77030-402 77030-404
(5-Fluoro-2-(trifluoromethyl)phenyl)acetic acid 98%
(5-Fluoro-2-(trifluoromethyl)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₆F₄O₂
MW: 222.14 g/mol
MDL Number: MFCD00236304
CAS Number: 239135-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR