5-Bromo-2-carboxy-3-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-methylpyridine-2-carboxylic acid, 5-Bromo-3-methylpicolinic acid

A208080-100G A208080-10G A208080-1G A208080-25G A208080-5G
77012-880EA 182.48 USD
77012-880 77012-882 77012-884 77012-886 77012-888
5-Bromo-2-carboxy-3-methylpyridine 98%
5-Bromo-2-carboxy-3-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
MDL Number: MFCD07375125
CAS Number: 886365-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR