Piperidine-4-carbonitrile hydrochloride 95%

Supplier: Ambeed
Danger

Synonyms: 4-Cyanopiperidine hydrochloride

A208025-100G A208025-10G A208025-1G A208025-5G A208025-25G
77012-872EA 196.27 USD
77012-872 77012-874 77012-876 77012-878 77683-591
Piperidine-4-carbonitrile hydrochloride 95%
Piperidine-4-carbonitrile hydrochloride

Precaución: For research use only.

Formula: C₆H₁₁ClN₂
MW: 146.62 g/mol
MDL Number: MFCD09037930
CAS Number: 240401-22-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR