3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
RoflumilastRelated Compound D, 3-(Cyclopropylmethoxy)-4-(difluoromethoxy)benzoic acid
A208015-10G
A208015-1G
A208015-250MG
A208015-5G
A208015-25G
77030-388EA
91.24
USD
77030-388
77030-390
77030-392
77030-394
77671-840
3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid 98%
3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₁₂H₁₂F₂O₄ MW: 258.221 g/mol |
MDL Number:
MFCD04621687 CAS Number: 162401-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...