(2R,3R,4S,5R,6R)-2-(Allyloxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol 98%

Supplier: Ambeed
Warning

A208013-100MG A208013-1G A208013-250MG
76986-628EA 9 USD
76986-628 76986-630 76986-632
(2R,3R,4S,5R,6R)-2-(Allyloxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol 98%
(2R,3R,4S,5R,6R)-2-(Allyloxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

Precaución: for Research Use Only

Formula: C₉H₁₆O₆
MDL Number: MFCD08274511
CAS Number: 2595-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR