4-(4-Phenylbutoxy)benzoic acid 97%

Supplier: Ambeed
Warning

A207985-100G A207985-10G A207985-1G A207985-5G A207985-25G
77030-380EA 26 USD
77030-380 77030-382 77030-384 77030-386 77667-748
4-(4-Phenylbutoxy)benzoic acid 97%
4-(4-Phenylbutoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₈O₃
MDL Number: MFCD07787608
CAS Number: 30131-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR