5-(Trifluoromethyl)-1H-pyrrolo[3,2-b]pyridine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A207973-100MG A207973-250MG
76986-622EA 195.21 USD
76986-622 76986-624
5-(Trifluoromethyl)-1H-pyrrolo[3,2-b]pyridine-2-carboxylic acid 95%
5-(Trifluoromethyl)-1H-pyrrolo[3,2-b]pyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅F₃N₂O₂
MW: 230.14 g/mol
MDL Number: MFCD13196665
CAS Number: 920979-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR