1-(5-Chloropyridin-2-yl)ethanone 98%

Supplier: Ambeed
Warning

A207927-10G A207927-1G A207927-250MG A207927-25G A207927-5G A207927-100G
76986-610EA 51 USD
76986-610 76986-612 76986-614 76986-616 76986-618 77885-505
1-(5-Chloropyridin-2-yl)ethanone 98%
1-(5-Chloropyridin-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₆ClNO
MDL Number: MFCD07375071
CAS Number: 94952-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR