4-Amino-2,6-dimethylphenol 96%

Supplier: Ambeed
Warning

A207869-250MG A207869-25G A207869-5G
76999-546EA 37 USD
76999-546 76999-548 77881-490
4-Amino-2,6-dimethylphenol 96%
4-Amino-2,6-dimethylphenol

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MDL Number: MFCD10039560
CAS Number: 15980-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR