Methyl 2-amino-5-cyanobenzoate 98%

Supplier: Ambeed
Warning

A207822-100MG A207822-10G A207822-1G A207822-250MG
77030-372EA 48.81 USD
77030-372 77030-374 77030-376 77030-378
Methyl 2-amino-5-cyanobenzoate 98%
Methyl 2-amino-5-cyanobenzoate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.172 g/mol
MDL Number: MFCD11041293
CAS Number: 159847-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR