Quinolin-3-ylmethanol 95%

Supplier: Ambeed
Warning

A207815-10G A207815-1G A207815-250MG A207815-5G
77030-364EA 741.57 USD
77030-364 77030-366 77030-368 77030-370
Quinolin-3-ylmethanol 95%
Quinolin-3-ylmethanol

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MDL Number: MFCD03412552
CAS Number: 13669-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR