8-Methylnaphthalen-1-amine 95%

Supplier: Ambeed
Warning

A207614-100MG A207614-250MG A207614-5G A207614-1G
77012-866EA 153.84 USD
77012-866 77012-868 77012-870 77683-588
8-Methylnaphthalen-1-amine 95%
8-Methylnaphthalen-1-amine

Precaución: For research use only.

Formula: C₁₁H₁₁N
MW: 157.21 g/mol
MDL Number: MFCD15523638
CAS Number: 130523-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR