Dimethyl 4,5-dihydroxyphthalate 97%

Supplier: Ambeed
Warning

A207572-100MG A207572-250MG
77012-862EA 75.33 USD
77012-862 77012-864
Dimethyl 4,5-dihydroxyphthalate 97%
Dimethyl 4,5-dihydroxyphthalate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₆
MW: 226.18 g/mol
MDL Number: MFCD20265697
CAS Number: 66323-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR