4-Chloro-7-methoxy-6-nitroquinazoline 95%

Supplier: Ambeed
Warning

A207372-100MG A207372-1G A207372-250MG A207372-5G
77030-348EA 64.72 USD
77030-348 77030-350 77030-352 77030-354
4-Chloro-7-methoxy-6-nitroquinazoline 95%
4-Chloro-7-methoxy-6-nitroquinazoline

Precaución: for Research Use Only

Formula: C₉H₆ClN₃O₃
MW: 239.62 g/mol
MDL Number: MFCD11869864
CAS Number: 55496-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR