3-Iodo-5-methyl-1H-pyrrolo[3,2-b]pyridine 95%

Supplier: Ambeed
Warning

A207342-100MG A207342-1G A207342-250MG
77012-846EA 16 USD
77012-846 77012-848 77012-850
3-Iodo-5-methyl-1H-pyrrolo[3,2-b]pyridine 95%
3-Iodo-5-methyl-1H-pyrrolo[3,2-b]pyridine

Precaución: for Research Use Only

Formula: C₈H₇IN₂
MDL Number: MFCD09880091
CAS Number: 1000343-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR