Dimethyl 2,5-dibromoisophthalate 96%

Supplier: Ambeed
Warning

A207206-100MG A207206-250MG
77012-842EA 21.22 USD
77012-842 77012-844
Dimethyl 2,5-dibromoisophthalate 96%
Dimethyl 2,5-dibromoisophthalate

Precaución: for Research Use Only

Formula: C₁₀H₈Br₂O₄
MW: 351.98 g/mol
MDL Number: MFCD27923417
CAS Number: 1337958-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR