2,4,5-Tribromothiazole 95%

Supplier: Ambeed
Warning

A207113-1G A207113-5G
76986-602EA 259.93 USD
76986-602 76986-604
2,4,5-Tribromothiazole 95%
2,4,5-Tribromothiazole

Precaución: for Research Use Only

Formula: C₃Br₃NS
MW: 321.81 g/mol
MDL Number: MFCD19543764
CAS Number: 57314-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR