tert-Butyl ((2S,3S)-4-chloro-3-hydroxy-1-phenylbutan-2-yl)carbamate 95%

Supplier: Ambeed
Warning

A206916-100G A206916-1G A206916-5G
77030-318EA 1397.21 USD
77030-318 77030-320 77030-322
tert-Butyl ((2S,3S)-4-chloro-3-hydroxy-1-phenylbutan-2-yl)carbamate 95%
tert-Butyl ((2S,3S)-4-chloro-3-hydroxy-1-phenylbutan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₅H₂₂ClNO₃
MW: 299.79 g/mol
MDL Number: MFCD02183480
CAS Number: 165727-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR