Fmoc-S-trityl-L-penicillamine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Pen(Trt)-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methyl-3-(tritylthio)butanoic acid

A206854-1G A206854-5G A206854-100MG A206854-250MG
76999-534EA 26 USD
76999-534 77550-326 77881-485 77881-486
Fmoc-S-trityl-L-penicillamine 97%
Fmoc-S-trityl-L-penicillamine

Precaución: for Research Use Only

Formula: C₃₉H₃₅NO₄S
MW: 613.772 g/mol
MDL Number: MFCD00237388
CAS Number: 201531-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR