7-Bromo-2,3-dihydrobenzofuran 98%

Supplier: Ambeed
Warning

A206773-1G A206773-250MG A206773-5G
77030-314EA 53.05 USD
77030-314 77030-316 77593-348
7-Bromo-2,3-dihydrobenzofuran 98%
7-Bromo-2,3-dihydrobenzofuran

Precaución: for Research Use Only

Formula: C₈H₇BrO
MW: 199.045 g/mol
MDL Number: MFCD09266210
CAS Number: 206347-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR