(1S,3aR,6aS)-tert-Butyl octahydrocyclopenta[c]pyrrole-1-carboxylate oxalate 95%

Supplier: Ambeed
Warning

A206503-100MG A206503-250MG
77030-310EA 23.34 USD
77030-310 77030-312
(1S,3aR,6aS)-tert-Butyl octahydrocyclopenta[c]pyrrole-1-carboxylate oxalate 95%
(1S,3aR,6aS)-tert-Butyl octahydrocyclopenta[c]pyrrole-1-carboxylate oxalate

Precaución: for Research Use Only

Formula: C₁₄H₂₃NO₆
MW: 301.3400 g/mol
MDL Number: MFCD22124391
CAS Number: 907606-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR