3-Chloro-4-ethynylaniline 95%

Supplier: Ambeed
Warning

A206456-1G A206456-250MG
77012-806EA 297 USD
77012-806 77012-808
3-Chloro-4-ethynylaniline 95%
3-Chloro-4-ethynylaniline

Precaución: for Research Use Only

Formula: C₈H₆ClN
MDL Number: MFCD11036466
CAS Number: 80690-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR