5-Bromo-2-(cyclopropylmethoxy)pyridine 98%
Supplier: Ambeed
|
Warning
|
A206355-1G
A206355-250MG
A206355-5G
77030-304EA
125.19
USD
77030-304
77030-306
77030-308
5-Bromo-2-(cyclopropylmethoxy)pyridine 98%
5-Bromo-2-(cyclopropylmethoxy)pyridine
Precaución: for Research Use Only
Formula:
C₉H₁₀BrNO MW: 228.08 g/mol |
MDL Number:
MFCD13190758 CAS Number: 494772-02-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...