Methyl 4-chloro-6-methoxypicolinate 95%

Supplier: Ambeed
Warning

A206198-100MG A206198-5G
77012-802EA 119.89 USD
77012-802 77012-804
Methyl 4-chloro-6-methoxypicolinate 95%
Methyl 4-chloro-6-methoxypicolinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₃
MW: 201.60 g/mol
MDL Number: MFCD18257705
CAS Number: 204378-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR