1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13-tetraoxa-4-azahexadecan-16-oic acid 97%
Supplier: Ambeed
|
Warning
|
A206129-100MG
A206129-10G
A206129-1G
A206129-250MG
A206129-25G
A206129-5G
76986-584EA
32.89
USD
76986-584
76986-586
76986-588
76986-590
76986-592
76986-594
1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13-tetraoxa-4-azahexadecan-16-oic acid 97%
1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13-tetraoxa-4-azahexadecan-16-oic acid
Precaución: for Research Use Only
Formula:
C₂₄H₂₉NO₇ MW: 443.48 g/mol |
MDL Number:
MFCD06656471 CAS Number: 867062-95-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...