4-Nitrobenzo[d]isoxazole 95%

Supplier: Ambeed
Warning

A206107-100MG A206107-1G A206107-250MG
77012-796EA 110 USD
77012-796 77012-798 77012-800
4-Nitrobenzo[d]isoxazole 95%
4-Nitrobenzo[d]isoxazole

Precaución: for Research Use Only

Formula: C₇H₄N₂O₃
MDL Number: MFCD26743746
CAS Number: 1823353-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR