3-(tert-Butyldimethylsilyloxy)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(tert-Butyl dimethylsiloxy)phenyl boronic acid, (3-((tert-Butyldimethylsilyl)oxy)phenyl)boronic acid, 3-(tert-Butyldimethylsiloxy)benzeneboronic acid

A206062-1G A206062-5G A206062-25G
76999-496EA 29 USD
76999-496 76999-498 77671-822
3-(tert-Butyldimethylsilyloxy)phenylboronic acid 98%
3-(tert-Butyldimethylsilyloxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁BO₃Si
MDL Number: MFCD03701505
CAS Number: 261621-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR