Ethyl-1H-pyrrolo[3,2-c]pyridine-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1H-pyrrolo[3,2-c]pyridine-2-carboxylate

A206024-1G A206024-250MG A206024-5G
77030-290EA 112.46 USD
77030-290 77030-292 77030-294
Ethyl-1H-pyrrolo[3,2-c]pyridine-2-carboxylate 97%
Ethyl-1H-pyrrolo[3,2-c]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.2000 g/mol
MDL Number: MFCD09878684
CAS Number: 800401-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR