Monomethyl-5-nitroisophthalate 95%

Supplier: Ambeed
Warning

Synonyms: 5-Nitroisophthalic acid monomethyl ester, Methyl 5-nitroisophthalate

A206002-100G A206002-25G A206002-500G A206002-5G
76999-488EA 45.62 USD
76999-488 76999-490 76999-492 76999-494
Monomethyl-5-nitroisophthalate 95%
Monomethyl-5-nitroisophthalate

Precaución: for Research Use Only

Formula: C₉H₇NO₆
MW: 225.16 g/mol
MDL Number: MFCD00009793
CAS Number: 1955-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR