[(2-Di-tert-butylphosphino-3,6-dimethoxy-2',4',6'-triisopropyl-1,1'-biphenyl)-2-(2'-amino-1,1'-biphenyl)]palladium(II) methanesulfonate 97%

Supplier: Ambeed
Warning

Synonyms: tBuBrettPhos Pd G3

A206000-100MG A206000-250MG
77012-792EA 16.98 USD
77012-792 77012-794
[(2-Di-tert-butylphosphino-3,6-dimethoxy-2',4',6'-triisopropyl-1,1'-biphenyl)-2-(2'-amino-1,1'-biphenyl)]palladium(II) methanesulfonate 97%
[(2-Di-tert-butylphosphino-3,6-dimethoxy-2',4',6'-triisopropyl-1,1'-biphenyl)-2-(2'-amino-1,1'-biphenyl)]palladium(II) methanesulfonate

Precaución: for Research Use Only

Formula: C₄₄H₆₂NO₅PPdS
MW: 854.43 g/mol
MDL Number: MFCD25976528
CAS Number: 1536473-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR