Isoxazol-4-ylmethanamine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A205897-100MG
A205897-250MG
77012-788EA
64
USD
77012-788
77012-790
Isoxazol-4-ylmethanamine hydrochloride 95%
Isoxazol-4-ylmethanamine hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₇ClN₂O |
MDL Number:
MFCD06738928 CAS Number: 173850-71-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...