2-Amino-6-methylphenol 95%

Supplier: Ambeed
Warning

A205732-10G A205732-1G A205732-250MG A205732-5G A205732-25G
76999-478EA 127 USD
76999-478 76999-480 76999-482 76999-484 77881-483
2-Amino-6-methylphenol 95%
2-Amino-6-methylphenol

Precaución: for Research Use Only

Formula: C₇H₉NO
MDL Number: MFCD03788432
CAS Number: 17672-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR