5-Amino-2,3-dihydroindole-1-carboxylic acid tert-butyl ester 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 5-amino-2,3-dihydro-1H-indole-1-carboxylate, tert-Butyl 5-aminoindoline-1-carboxylate

A205563-100MG A205563-250MG A205563-1G A205563-5G
76986-558EA 30.77 USD
76986-558 76986-560 77593-338 77612-014
5-Amino-2,3-dihydroindole-1-carboxylic acid tert-butyl ester 95%
5-Amino-2,3-dihydroindole-1-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₂
MW: 234.3 g/mol
MDL Number: MFCD08059270
CAS Number: 129487-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR