Decoquinate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylate, Ethyl 6-(decyloxy)-7-ethoxy-4-hydroxyquinoline-3-carboxylate, 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylic Acid Ethyl Ester

A205365-100G A205365-100MG A205365-25G A205365-500G
76999-470EA 113.52 USD
76999-470 76999-472 76999-474 76999-476
Decoquinate 98%
Decoquinate

Precaución: for Research Use Only

Formula: C₂₄H₃₅NO₅
MW: 417.55 g/mol
MDL Number: MFCD00673686
CAS Number: 18507-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR