2-Chloro-6-fluoro-3-methoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

A205252-100MG A205252-1G A205252-250MG A205252-5G
76999-462EA 5.31 USD
76999-462 76999-464 76999-466 76999-468
2-Chloro-6-fluoro-3-methoxyphenylboronic acid 97%
2-Chloro-6-fluoro-3-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BClFO₃
MW: 204.39 g/mol
MDL Number: MFCD09972096
CAS Number: 1072945-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR