1-(6-Aminohexyl)-1H-pyrrole-2,5-dione 2,2,2-trifluoroacetate 97%

Supplier: Ambeed
Warning

A205242-100MG A205242-1G A205242-250MG A205242-5G
77030-284EA 23 USD
77030-284 77030-286 77593-336 77671-814
1-(6-Aminohexyl)-1H-pyrrole-2,5-dione 2,2,2-trifluoroacetate 97%
1-(6-Aminohexyl)-1H-pyrrole-2,5-dione 2,2,2-trifluoroacetate

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₂·CF₃CO₂H
MDL Number: MFCD11519191
CAS Number: 731862-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR