Tyrphostin AG 490 98%

Supplier: Ambeed
Danger

A205043-10MG A205043-1MG A205043-250MG A205043-50MG A205043-100MG
77012-774EA 22 USD
77012-774 77012-776 77012-778 77012-780 77890-235
Tyrphostin AG 490 98%
Tyrphostin AG 490

Precaución: for Research Use Only

Formula: C₁₇H₁₄N₂O₃
MDL Number: MFCD00236452
CAS Number: 133550-30-8
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR