2-Chloro-4-cyano-6-fluorobenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Carboxy-3-chloro-5-fluorobenzonitrile

A204779-100MG A204779-1G A204779-250MG
77012-768EA 71.09 USD
77012-768 77012-770 77012-772
2-Chloro-4-cyano-6-fluorobenzoic acid 97%
2-Chloro-4-cyano-6-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₃ClFNO₂
MW: 199.566 g/mol
MDL Number: MFCD22690817
CAS Number: 1258298-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR