(S)-(-)-N-Benzyl-α-methylbenzylamine 97%

Supplier: Ambeed
Warning

Synonyms: S-(-)-N-Benzyl-1-phenylethylamine, (S)-(-)-N-Benzyl-1-phenylethylamine

A204708-100G A204708-10G A204708-1G A204708-25G A204708-500G A204708-5G
76999-450EA 58.35 USD
76999-450 76999-452 76999-454 76999-456 76999-458 76999-460
(S)-(-)-N-Benzyl-α-methylbenzylamine 97%
(S)-(-)-N-Benzyl-α-methylbenzylamine

Precaución: for Research Use Only

Formula: C₁₅H₁₇N
MW: 211.31 g/mol
MDL Number: MFCD00066325
CAS Number: 17480-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR