Ethyl 2-(2-hydroxyphenoxy)acetate 97%

Supplier: Ambeed
Warning

A204616-250MG A204616-25G A204616-5G A204616-1G
77012-756EA 20 USD
77012-756 77012-758 77012-760 77890-234
Ethyl 2-(2-hydroxyphenoxy)acetate 97%
Ethyl 2-(2-hydroxyphenoxy)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MDL Number: MFCD00547114
CAS Number: 99186-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR