3-Bromo-5-fluoro-pyridine-4-carboxylic acid 96%

Supplier: Ambeed
Warning

A204591-100MG A204591-1G A204591-250MG A204591-5G
77030-260EA 9 USD
77030-260 77030-262 77030-264 77030-266
3-Bromo-5-fluoro-pyridine-4-carboxylic acid 96%
3-Bromo-5-fluoro-pyridine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₂
MDL Number: MFCD13181633
CAS Number: 955372-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR