3,4-Dihydroxybenzophenone 98%

Supplier: Ambeed
Warning

Synonyms: (3,4-Dihydroxyphenyl)(phenyl)methanone

A204498-100G A204498-10G A204498-5G A204498-25G
77030-246EA 93.36 USD
77030-246 77030-248 77030-250 77671-808
3,4-Dihydroxybenzophenone 98%
3,4-Dihydroxybenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₃
MW: 214.2200 g/mol
MDL Number: MFCD00477203
CAS Number: 10425-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR