1,1'-(Bicyclo[1.1.1]pentane-1,3-diyl)diethanone 95%

Supplier: Ambeed
Warning

Synonyms: 1-(3-Acetyl-1-bicyclo[1.1.1]pentanyl)ethanone

A204444-100MG A204444-1G A204444-250MG
77012-750EA 24.41 USD
77012-750 77012-752 77012-754
1,1'-(Bicyclo[1.1.1]pentane-1,3-diyl)diethanone 95%
1,1'-(Bicyclo[1.1.1]pentane-1,3-diyl)diethanone

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
MDL Number: MFCD23164149
CAS Number: 115913-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR