2-Aminopyrimidine-5-carbonitrile 97%

Supplier: Ambeed
Warning

A204264-1G A204264-250MG A204264-5G A204264-10G
77012-744EA 25.47 USD
77012-744 77012-746 77012-748 77683-574
2-Aminopyrimidine-5-carbonitrile 97%
2-Aminopyrimidine-5-carbonitrile

Precaución: For research use only.

Formula: C₅H₄N₄
MW: 120.1121 g/mol
MDL Number: MFCD00232791
CAS Number: 1753-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR