Pyrazine-2,5-diamine 97%

Supplier: Ambeed
Warning

A204194-100MG A204194-250MG A204194-5G A204194-1G
77030-234EA 20 USD
77030-234 77030-236 77030-238 77593-330
Pyrazine-2,5-diamine 97%
Pyrazine-2,5-diamine

Precaución: for Research Use Only

Formula: C4H6N4
MDL Number: MFCD09835012
CAS Number: 768386-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR